missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford D-Cycloserine
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine.
D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine. It inhibits cell wall synthesis in certain bacteria, including Mycobacterium tuberculosis. While it was initially identified as an antibiotic, D-cycloserine has gained attention for its activity as a partial agonist at the N-methyl-D-aspartate (NMDA) receptor, a type of glutamate receptor in the brain. Researchers study the mechanisms of neurotransmission and synaptic plasticity due to its interactions with NMDA receptors.
Specifications
Specifications
| Chemical Name or Material | D-Cycloserine |
| CAS | 68-41-7 |
| Quantity | 1 g |
| Molecular Formula | C3H6N2O2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction