missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Carvone, 98%, synthetic

Product Code. p-3523412 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10135922

Brand: Thermo Scientific Acros 377840250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 2244-16-8 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2244-16-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00062997
InChI Key ULDHMXUKGWMISQ-VIFPVBQESA-N
Synonym d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
PubChem CID 16724
ChEBI CHEBI:15399
IUPAC Name (5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC1=CCC(CC1=O)C(=C)C

Specifications

Color Colorless to Yellow
Density 0.9560g/mL
Boiling Point 230.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5010
Quantity 25 mL
Specific Gravity 0.956
Specific Rotation Condition + 57.00 (20.00°C c=neat)
Specific Rotation + 57.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol
Formula Weight 150.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material D(+)-Carvone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.