missing translation for 'onlineSavingsMsg'
Learn More

D-Carnitine, TRC

Product Code. 30137088
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137088 50 mg 50mg
30136686 1 g 1g
30011064 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137088 Supplier Toronto Research Chemicals Supplier No. TRCC18410550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-14-0
Molecular Formula C7 H15 N O3
Molecular Weight (g/mol) 161.2
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, inner salt, (2S)-, 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, inner salt, (S)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, hydroxide, inner salt, D- (8CI), (+)-Carnitine, (+)-D-Carnitine, D-(+)-Carnitine, D-Carnitine, d-Carnitine
IUPAC Name (3S)-3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)C[C@@H](O)CC(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m0/s1
Formula Weight 161.1052
Percent Purity >95%
Chemical Name or Material D-Carnitine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.