missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Camphor, 97%

Product Code. 10135240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135240 100 g Plastic bottle 100g
10637172 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10135240 Supplier Thermo Scientific Acros Supplier No. 227841000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 464-49-3 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-49-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00064149,MFCD00074738,MFCD00064149,MFCD00064149
InChI Key DSSYKIVIOFKYAU-XVKPBYJWSA-N
Synonym +-camphor, d-camphor, 1r-+-camphor, r-camphor, +-bornan-2-one, r-+-camphor, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, 1r,4r-camphor, camphor d, +-2-bornanone
PubChem CID 159055
ChEBI CHEBI:15396
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)[C@H]2CC[C@@]1(C)C(=O)C2

Specifications

Melting Point 176.0°C to 180.0°C
Color White
Boiling Point 204.0°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Fieser 13,61
Merck Index 15, 1734
Specific Rotation + 42.50
Solubility Information Solubility in water: 1g/800mL water (25°C) (giving a colloidal solutio. Other solubilities: at 25°C 1 g dissolves in 1 mL alcohol,1 mL ether,,0.5 mL chloroform,0.4 mL benzene,0.4 mL acetone,,1.5 mL turpentine oil,0.5 mL glacial acetic acid
Formula Weight 152.24
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material D(+)-Camphor, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • D(+)-Camphor
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.