missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Calcium pantothenate, 98%

Product Code. 10439871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10439871 100 g Plastic Bottle 100g
10358270 5 g Glass Bottle 5g
10712964 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10439871 Supplier Thermo Scientific Chemicals Supplier No. 243301000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-08-6 | C18H32CaN2O10 | 476.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

Melting Point 190°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 8.2 to 8.6% (Ca) On dried substance (Complexometry)
Loss on Drying 5.0% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula [HOCH2C(CH3)2CHOHCONHCH2CH2COO]2Ca
Specific Rotation + 25
Merck Index 15, 7118
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: soluble in glycerine,very slightly soluble in ethanol,insoluble in ether,chloroform
Quantity 100 g
Formula Weight 476.52
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Calcium pantothenate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.