missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BLNK Monoclonal specifically detects BLNK in Human samples. It is validated for ELISA.
Spécification
Spécification
| Antigen | BLNK |
| Gene Alias | AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human BLNK (Accession#: AAH18906.1; Met 1-Ser 456). |
| Primary or Secondary | Primary |
| Content And Storage | Store at 4C in the dark. |
| Product Type | Antibody |
| Isotype | IgG |
| Product Line | Novus |
Nom du produit
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?