missing translation for 'onlineSavingsMsg'
Learn More

D-Beta-Imidazole lactic Acid Monohydrate, TRC

Product Code. 30240884
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240884 100 mg 100mg
30240491 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240884 Supplier Toronto Research Chemicals Supplier No. TRCI354990100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246814-96-9
Molecular Formula C6 H8 N2 O3 . H2 O
Molecular Weight (g/mol) 174.15
IUPAC Name (2R)-2-hydroxy-3-(1H-imidazol-4-yl)propanoic acid;hydrate
SMILES O.O[C@H](Cc1c[nH]cn1)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H8N2O3.H2O/c9-5(6(10)11)1-4-2-7-3-8-4;/h2-3,5,9H,1H2,(H,7,8)(H,10,11);1H2/t5-;/m1./s1
Formula Weight 174.0641
Chemical Name or Material D-beta-Imidazolelactic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.