missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Aspartic acid, 99+%

Product Code. 10145403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10145403 100 g Glass Bottle 100g
10574274 5 g Glass Bottle 5g
10296540 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10145403 Supplier Thermo Scientific Chemicals Supplier No. 204261000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1783-96-6 | C4H7NO4 | 133.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1783-96-6
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00063081
InChI Key CKLJMWTZIZZHCS-UWTATZPHSA-N
Synonym d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
PubChem CID 83887
ChEBI CHEBI:17364
IUPAC Name (2R)-2-aminobutanedioic acid
SMILES C(C(C(=O)O)N)C(=O)O

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.4% max. (105°C, 3 hrs)
Other Amino Acids 0.3% max. (TLC)
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CO2H
Residue after Ignition 0.1% max.
Beilstein 04, 471
Specific Rotation − 25.80
Merck Index 15, 830
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether
Specific Rotation Condition − 25.80 (20.00°C c=5,5N HCl)
Quantity 100 g
Formula Weight 133.1
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Aspartic acid, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.