missing translation for 'onlineSavingsMsg'
Learn More

D-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific™

Product Code. 15400689
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15400689

Brand: Thermo Scientific Alfa Aesar H63795.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64960-75-4
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00077101
InChI Key MXWMFBYWXMXRPD-UHFFFAOYNA-N
Synonym h-d-asp otbu-oh, d-aspartic acid 4-tert-butyl ester, r-2-amino-4-tert-butoxy-4-oxobutanoic acid, 2r-2-amino-4-tert-butoxy-4-oxobutanoic acid, d-aspartic acid beta-tert-butyl ester, d-aspartic acid 4-1,1-dimethylethyl ester, 2r-2-amino-4-tert-butoxy-4-oxo-butanoic acid, 2r-2-amino-4-2-methylpropan-2-yl oxy-4-oxobutanoic acid, 2r-2-azanyl-4-2-methylpropan-2-yl oxy-4-oxidanylidene-butanoic acid
PubChem CID 7018828
SMILES CC(C)(C)OC(=O)CC(N)C(O)=O

Specifications

IUPAC Name 2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Quantity 250 mg
Formula Weight 189.21
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Aspartic acid 4-tert-butyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.