missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Asparagine Monohydrate, TRC

Product Code. 30344667
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344667 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344667 Supplier Toronto Research Chemicals Supplier No. TRCA780125250MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-(-)-Asparagine Monohydrate
CAS 5794-24-1
Molecular Formula C4H8N2O3 . H2O
Quantity 250 mg
Synonym D-Asparagine, hydrate (1:1), Asparagine, monohydrate, D- (8CI), D-Asparagine, monohydrate (9CI), (R)-2,4-Diamino-4-oxobutanoic acid hydrate, D-Asparagine monohydrate
InChI Formula InChI=1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m1./s1
SMILES O.N[C@H](CC(=O)N)C(=O)O
IUPAC Name (2R)-2,4-diamino-4-oxobutanoic acid;hydrate
Molecular Weight (g/mol) 150.13
Formula Weight 150.0641
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.