missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Asparagine monohydrate

Product Code. 17281136 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17281136 5 g 5g
17271136 1 g 1g
17291136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17281136 Supplier Thermo Scientific Chemicals Supplier No. 204270050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5794-24-1 | C4H10N2O4 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-24-1
Molecular Weight (g/mol) 150.13
InChI Key RBMGJIZCEWRQES-UHFFFAOYNA-N
IUPAC Name 2-amino-3-carbamoylpropanoic acid hydrate
SMILES O.NC(CC(N)=O)C(O)=O

Specifications

Melting Point 280.0°C
Molecular Formula C4H10N2O4
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Beilstein 04, 471
Merck Index 14, 837
Solubility Information Solubility aq. soln.: 35 g/L (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene
Quantity 5 g
Formula Weight 150.13
Chemical Name or Material D-Asparagine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.