missing translation for 'onlineSavingsMsg'
Learn More

D-Arginine methyl ester dihydrochloride, 98%, Thermo Scientific™

Catalog Number 15469178
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
78851-84-0
C7H18Cl2N4O2
261.147
MFCD00153417
XQYZOBNLCUAXLF-ZJIMSODOSA-N
h-d-arg-ome.2hcl, h-d-arg-ome 2hcl, r-methyl 2-amino-5-guanidinopentanoate dihydrochloride, d-arginine methyl ester dihydrochloride, d-arginine methyl ester di hydrochloride, d-arginine, methyl ester, dihydrochloride, methyl 2r-2-amino-5-carbamimidamidopentanoate dihydrochloride, h-d-arg-ome 2 hcl, h-d-arg-ome. 2hcl, d-arginine methyl ester dihydrochlorid
12714066
methyl (2R)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
COC(=O)C(CCCN=C(N)N)N.Cl.Cl
This item is not returnable. View return policy
78851-84-0
C7H18Cl2N4O2
261.147
MFCD00153417
XQYZOBNLCUAXLF-ZJIMSODOSA-N
h-d-arg-ome.2hcl, h-d-arg-ome 2hcl, r-methyl 2-amino-5-guanidinopentanoate dihydrochloride, d-arginine methyl ester dihydrochloride, d-arginine methyl ester di hydrochloride, d-arginine, methyl ester, dihydrochloride, methyl 2r-2-amino-5-carbamimidamidopentanoate dihydrochloride, h-d-arg-ome 2 hcl, h-d-arg-ome. 2hcl, d-arginine methyl ester dihydrochlorid
12714066
methyl (2R)-2-amino-5-(diaminomethylideneamino)pentanoate;dihydrochloride
COC(=O)C(CCCN=C(N)N)N.Cl.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 261.15
Percent Purity 98%
Quantity 1 g
Sensitivity Hygroscopic
Physical Form Solid
Chemical Name or Material D-Arginine methyl ester dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'