missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Arabitol, 99%

Product Code. 10147513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147513 25 g Glass bottle 25g
10702881 5 g Glass bottle 5g
10167893 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147513 Supplier Thermo Scientific Chemicals Supplier No. 302870250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-82-4 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-82-4
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004709
InChI Key HEBKCHPVOIAQTA-QWWZWVQMSA-N
Synonym d-arabinitol, arabitol, dl-arabitol, d-arabitol, d-+-arabitol, d-+-arabinitol, d-lyxitol, lyxitol, dl-arabinitol, d-arabinol
PubChem CID 94154
ChEBI CHEBI:18333
IUPAC Name (2R,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Melting Point 97°C to 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition + 12.00 (20.00°C c=9,borax solution)
Specific Rotation + 12
Formula Weight 152.15
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material D-Arabitol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.