missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress D-Arabitol

Product Code. 16442599
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442599 Supplier MedChemExpress Supplier No. HYN3686
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress D-Arabitol is a polyol and its accumulation may cause a neurotoxic effect in human.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 152.15 g/mol
Shipping Condition Room temperature
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 488-82-4
Product Type Natural Products
Purity 97.0%
Solubility DMSO : 100 mg/mL (657.25 mM∣Need ultrasonic)
Target Endogenous Metabolite
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.