missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Arabinose (White Powder), Fisher BioReagents

Product Code. 10216054
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10216054 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10216054 Supplier Fisher BioReagents Supplier No. BP2504100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
D-(-)-Arabinose is a component used for culture media.
TRUSTED_SUSTAINABILITY

Specifications

CAS 28697-53-2
Melting Point 152°C
Color White
Assay Percent Range ≥99 %
Packaging Amber Glass
Molecular Formula C5H10O5
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
Quantity 100 g
Specific Rotation -107 to -97° (+ or -)
Synonym alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
SMILES OCC(O)[C@@H](O)[C@H](O)C=O
IUPAC Name (4S,5R)-oxane-2,3,4,5-tetrol
Molecular Weight (g/mol) 150.13
PubChem CID 71313478
Formula Weight 150.07
Percent Purity 99% min.
Physical Form Powder or Solid
Chemical Name or Material D-(-)-Arabinose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.