missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Arabinitol, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | D-(+)-Arabitol |
| CAS | 488-82-4 |
| Molecular Formula | C5 H12 O5 |
| Quantity | 50 g |
| Synonym | Arabinitol, D- (8CI), (+)-Arabitol, D-(+)-Arabitol, D-Arabitol, D-Lyxitol, D-arabino-Pentitol, D-lyxo-Pentitol, NSC 25288 |
| InChI Formula | InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m1/s1 |
| SMILES | OC[C@@H](O)C(O)[C@H](O)CO |
| IUPAC Name | (2R,4R)-pentane-1,2,3,4,5-pentol |
| Molecular Weight (g/mol) | 152.15 |
| Formula Weight | 152.0685 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction