missing translation for 'onlineSavingsMsg'
Learn More

D-AP5, MedChemExpress

Product Code. 16473128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473128 10 mg 10mg
16415867 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16473128 Supplier Medchem Express Supplier No. HY100714A
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-AP5 (D-APV) is a selective and competitive NMDA receptor antagonist with a Kd of 1.4 μM. D-AP5 (D-APV) inhibits the glutamate binding site of NMDA receptors.

D-AP5 (D-APV) is a selective and competitive NMDA receptor antagonist with a Kd of 1.4 μM. D-AP5 (D-APV) inhibits the glutamate binding site of NMDA receptors.

Specifications

Chemical Name or Material D-AP5
Physical Form Solid
CAS 79055-68-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C5H12NO5P
For Use With (Application) Neuroscience-Neurodegeneration
Synonym D-APV D-2-Amino-5-phosphonovaleric acid
Solubility Information H2O : 27.78 mg/mL (140.92 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES N[C@H](CCCP(O)(O)=O)C(O)=O
Molecular Weight (g/mol) 197.13
Formula Weight 197.13
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.