missing translation for 'onlineSavingsMsg'
Learn More

D-AP5, MedChemExpress

Product Code. 16415867
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16415867

Brand: Medchem Express HY100714A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-AP5 (D-APV) is a selective and competitive NMDA receptor antagonist with a Kd of 1.4 μM. D-AP5 (D-APV) inhibits the glutamate binding site of NMDA receptors.

D-AP5 (D-APV) is a selective and competitive NMDA receptor antagonist with a Kd of 1.4 μM. D-AP5 (D-APV) inhibits the glutamate binding site of NMDA receptors.

Specifications

Chemical Name or Material D-AP5
Physical Form Solid
CAS 79055-68-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C5H12NO5P
For Use With (Application) Neuroscience-Neurodegeneration
Synonym D-APV D-2-Amino-5-phosphonovaleric acid
Solubility Information H2O : 27.78 mg/mL (140.92 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES N[C@H](CCCP(O)(O)=O)C(O)=O
Molecular Weight (g/mol) 197.13
Formula Weight 197.13
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.