missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Alpha-Bromocamphor-8-sulfonic Acid Ammonium Salt, TRC

Product Code. 30341516
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341516 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341516 Supplier Toronto Research Chemicals Supplier No. TRCB68209010G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-(+)-a-Bromocamphor-8-sulfonic Acid Ammonium Salt
CAS 14575-84-9
Molecular Formula C10 H14 Br O4 S . H4 N
Quantity 10 g
Synonym Bicyclo[2.2.1]heptane-7-methanesulfonic acid, 2-bromo-4,7-dimethyl-3-oxo-, ammonium salt (1:1), (1S,2S,4R,7R)-, 8-Bornanesulfonic acid, 3-bromo-2-oxo-, ammonium salt, (+)- (8CI), Bicyclo[2.2.1]heptane-7-methanesulfonic acid, 3-bromo-1,7-dimethyl-2-oxo-, ammonium salt, (1R,3S,4S,7R)- (9CI), Bicyclo[2.2.1]heptane-7-methanesulfonic acid, 3-bromo-1,7-dimethyl-2-oxo-, ammonium salt, [1R-(endo,anti)]-, (1R)-(+)-3-Bromocamphor-8-sulfonic acid ammonium salt, Ammonium (+)-α-bromocamphor-π-sulfonate, Ammonium (1R)-(+)-3-bromocamphor-8-sulfonate, Ammonium [(1R)-(endo,anti)]-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptane-7-methanesulfonate, Ammonium d-α-bromocamphor-π-sulfonate, D-(+)-α-Bromocamphor-8-sulfonic Acid Ammonium Salt
InChI Formula InChI=1S/C10H15BrO4S.H3N/c1-9-4-3-6(7(11)8(9)12)10(9,2)5-16(13,14)15;/h6-7H,3-5H2,1-2H3,(H,13,14,15);1H3/t6-,7+,9+,10-;/m1./s1
SMILES [NH4+].C[C@@]1(CS(=O)(=O)[O-])[C@@H]2CC[C@@]1(C)C(=O)[C@H]2Br
IUPAC Name azanium;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonate
Molecular Weight (g/mol) 328.22
Formula Weight 327.014
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.