missing translation for 'onlineSavingsMsg'
Learn More

D-allo-Isoleucine, 97%, Thermo Scientific Chemicals

Catalog Number p-7029696
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1509-35-9
C6H13NO2
131.18
MFCD00066445
AGPKZVBTJJNPAG-CRCLSJGQSA-N
d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
94206
CHEBI:20899
(2R,3S)-2-amino-3-methylpentanoic acid
CC[C@H](C)[C@@H](N)C(O)=O
This item is not returnable. View return policy
1509-35-9
C6H13NO2
131.18
MFCD00066445
AGPKZVBTJJNPAG-CRCLSJGQSA-N
d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
94206
CHEBI:20899
(2R,3S)-2-amino-3-methylpentanoic acid
CC[C@H](C)[C@@H](N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽285°C (decomposition)
Beilstein 1721794
Formula Weight 131.18
Percent Purity 97%
Quantity 250 mg
Optical Rotation −37° (c=1 in 5N HCl)
Chemical Name or Material D-allo-Isoleucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'