missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine, TRC

Product Code. 30179211
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179211 25 g 25g
30179372 250 g 250g
30179314 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179211 Supplier Toronto Research Chemicals Supplier No. TRCA48099525G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338-69-2
Molecular Formula C3 H7 N O2
Molecular Weight (g/mol) 89.09
Synonym D-Alanine, (2R)-2-(Amino)propanoic Acid, (R)-2-Aminopropanoic Acid, (R)-Alanine, 2: PN: RU2506269 PAGE: 13 Claimed Sequence, Ba 2776, D(-)-α-Alanine, D-(-)-Alanine, D-α-Alanine, NSC 158286
IUPAC Name (2R)-2-aminopropanoic acid
SMILES C[C@@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1
Quantity 25 g
Formula Weight 89.0477
Shipping Condition Room Temperature
Chemical Name or Material D-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.