missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine Methyl Ester Hydrochloride, TRC

Product Code. 30309330
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309330 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309330 Supplier Toronto Research Chemicals Supplier No. TRCA48100510G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Alanine Methyl Ester Hydrochloride
CAS 14316-06-4
Molecular Formula C4H10ClNO2
Quantity 10 g
Synonym (R)-1-(Methoxycarbonyl)ethylamine Hydrochloride, (R)-2-Amino-propionic Acid Methyl Ester Hydrochloride, (R)-2-Aminopropionic Acid Methyl Ester Hydrochloride, (R)-Alanine Methyl Ester Hydrochloride, Methyl (R)-2-Aminopropanoate Hydrochloride, Methyl D-Alaninate Hydrochloride, NSC 523192
InChI Formula InChI=1S/C4H9NO2.ClH/c1-3(5)4(6)7-2;/h3H,5H2,1-2H3;1H/t3-;/m1./s1
SMILES Cl.COC(=O)[C@@H](C)N
IUPAC Name methyl (2R)-2-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 139.58
Formula Weight 139.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.