missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine ethyl ester hydrochloride, 95%

Product Code. 15429268
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429268 5 g 5g
15439268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429268 Supplier Thermo Scientific Alfa Aesar Supplier No. H62791.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6331-09-5 | C8H19NO2 | 161.25 g/mol

D-Alanine ethyl ester hydrochloride is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine ethyl ester hydrochloride is used as organic intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6331-09-5
MDL Number MFCD00190725

Specifications

Molecular Formula C5H11ClNO2
Synonym d-alanine ethyl ester hydrochloride, r-ethyl 2-aminopropanoate hydrochloride, h-d-ala-oet hcl, r-ethyl 2-amino-propionate hcl, ethyl 2r-2-aminopropanoate hydrochloride, h-ala-oblz. tos, h-d-ala-oet. hcl, 3-alanine ethyl ester hydrochloride, r-ethyl 2-amino-propionate hydrochloride
InChI Key CYHRNDXVTAWKMO-UHFFFAOYNA-N
SMILES CCOC(=O)C(C)N.Cl
IUPAC Name ethyl (2R)-2-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 152.60
Quantity 5 g
PubChem CID 10236019
Formula Weight 153.61
Percent Purity 95%
Chemical Name or Material D-Alanine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.