missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine benzyl ester p-toluenesulfonate, 98%, Thermo Scientific™

Product Code. 15438918
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438918 5 g 5g
15448918 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438918 Supplier Thermo Scientific Alfa Aesar Supplier No. H62412.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41036-32-2
Molecular Formula C17H21NO5S
Molecular Weight (g/mol) 351.42
MDL Number MFCD00039092
InChI Key NWOPHJSSBMABBD-DDWIOCJRSA-N
Synonym h-d-ala-obzl.tosoh, d-alanine benzyl ester p-toluenesulfonate salt, d-alanine benzyl ester p-toluenesulfonate, d-alanine benzyl ester tosylate, h-d-ala-obzl p-tosylate, h-d-ala-obzl.tos-oh, o-benzyl-d-alanine toluene-p-sulphonate, r-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2r-2-aminopropanoate; para-toluene sulfonate, d-ala-obzltosoh
PubChem CID 2802425
IUPAC Name 4-methylbenzene-1-sulfonic acid; benzyl (2R)-2-aminopropanoate
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.C[C@@H](N)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 351.42
Percent Purity 98%
Chemical Name or Material D-Alanine benzyl ester p-toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.