missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Alanine, 99%

Product Code. 11403933
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403933 100 g 100g
11483923 5 g 5g
11493923 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403933 Supplier Thermo Scientific Chemicals Supplier No. A10231.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol

D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.

Solubility
Soluble in water (155 mg/ml at 20°C), ethanol (slightly), and pyridine (slightly). Insoluble in ether, and acetone.

Notes
Store in cool, dry conditions in well sealed containers. Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338-69-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008077
InChI Key QNAYBMKLOCPYGJ-UWTATZPHSA-N
Synonym d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
PubChem CID 71080
ChEBI CHEBI:15570
IUPAC Name (2R)-2-aminopropanoic acid
SMILES C[C@@H](N)C(O)=O

Specifications

Melting Point ∼295°C (decomposition)
Beilstein 1720249
Merck Index 14,204
Solubility Information Soluble in water (155mg/ml at 20°C),ethanol (slightly),and pyridine (slightly). Insoluble in ether,and acetone.
Quantity 100 g
Formula Weight 89.09
Optical Rotation −14° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-Alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.