missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Alanine, 99+%

Product Code. 11329711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11329711 100 g Glass Bottle 100g
10551621 5 g Glass Bottle 5g
10561621 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329711 Supplier Thermo Scientific Chemicals Supplier No. 172081000
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338-69-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008077
InChI Key QNAYBMKLOCPYGJ-UWTATZPHSA-N
Synonym d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
PubChem CID 71080
ChEBI CHEBI:15570
IUPAC Name (2R)-2-aminopropanoic acid
SMILES C[C@@H](N)C(O)=O

Specifications

Melting Point 278.0°C to 282.0°C
Arsenic (As) 2ppm max. (As2O3)
Color White
Chloride 0.1% max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.2% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula CH3CH(NH2)CO2H
Specific Rotation − 14.50
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: 155g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
Specific Rotation Condition − 14.50 (20.00°C c=10,6N HCl)
Quantity 100 g
Formula Weight 89.09
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Alanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.