missing translation for 'onlineSavingsMsg'
Learn More

D-64131, MedChemExpress

Product Code. 16442657
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442657 50 mg 50mg
16426568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442657 Supplier Medchem Express Supplier No. HY15482

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-64131 is an orally active tubulin inhibitor, with an IC50 of 0.53 μM for tubulin polymerization. D-64131 has antimitotic activity. D-64131 can be used for cancer research.

D-64131 is an orally active tubulin inhibitor, with an IC50 of 0.53 μM for tubulin polymerization. D-64131 has antimitotic activity. D-64131 can be used for cancer research.

Specifications

Chemical Name or Material D-64131
CAS 74588-78-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13NO2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (397.96 mM)
SMILES O=C(C(N1)=CC2=C1C=CC(OC)=C2)C3=CC=CC=C3
Molecular Weight (g/mol) 251.28
Formula Weight 251.28
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.