missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-4-Hydroxyphenylglycine, 98+%

Product Code. 10731861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10731861 100 g Plastic bottle 100g
10042500 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10731861 Supplier Thermo Scientific Chemicals Supplier No. 201321000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22818-40-2 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22818-40-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00004262
InChI Key LJCWONGJFPCTTL-SSDOTTSWSA-N
Synonym d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
PubChem CID 89853
ChEBI CHEBI:15695
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)O

Specifications

Melting Point 240°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HOC6H4CH(NH2)CO2H
Quantity 100 g
Beilstein 14, I, 659
Specific Rotation Condition −156° (20°C c=1,1 N HCl)
Specific Rotation -156
Solubility Information Solubility in water: 5g/L (20 c)
Formula Weight 167.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material D(-)-4-Hydroxyphenylglycine, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • D(-)-4-Hydroxyphenylglycine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.