missing translation for 'onlineSavingsMsg'
Learn More

D-3263 hydrochloride, MedChemExpress

Product Code. 16483047
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483047 50 mg 50mg
16479537 5 mg 5mg
16451599 1 mg 1mg
16427066 100 mg 100mg
16426998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483047 Supplier Medchem Express Supplier No. HY16162A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-3263 hydrochloride is an enteric-coated, orally bioavailable (transient receptor potential melastatin member 8) TRPM8 agonist.

D-3263 hydrochloride is an enteric-coated, orally bioavailable (transient receptor potential melastatin member 8) TRPM8 agonist.

Specifications

Chemical Name or Material D-3263 hydrochloride
CAS 1008763-54-9
Color White
Purity Grade Notes Research
Molecular Formula C21H32ClN3O3
Quantity 50 mg
Solubility Information DMSO : 140 mg/mL (341.51 mM; Need ultrasonic)
SMILES [H]Cl.O=C1N(CCN)C2=CC(OC)=CC=C2N1C([C@H]3[C@H](C(C)C)CC[C@@H](C)C3)=O
Molecular Weight (g/mol) 409.95
Formula Weight 409.95
Percent Purity 97.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.