missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-3-Phenyllactic acid, 98%

Product Code. 15457467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457467 5 g 5g
15447467 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457467 Supplier Thermo Scientific Alfa Aesar Supplier No. H35096.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7326-19-4 | C9H10O3 | 166.18 g/mol

D-(+)-3-Phenyllactic acid is a chiral building block that is used in preparation of statine. Starting material in preparation of the hypoglycemic agent enlitazone and of (15)N-labeled phenylalanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-3-Phenyllactic acid is a chiral building block that is used in preparation of statine. Starting material in preparation of the hypoglycemic agent enlitazone and of (15)N-labeled phenylalanine.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7326-19-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00078062
InChI Key VOXXWSYKYCBWHO-MRVPVSSYSA-N
Synonym r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-3-phenyllactic acid, r-, a-hydroxy-3-phenylpropionic acid
PubChem CID 643327
ChEBI CHEBI:32978
IUPAC Name (2R)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 122°C to 127°C
Quantity 5 g
Solubility Information Soluble in water.
Optical Rotation +22° (c=1 in Water)
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material D-(+)-3-Phenyllactic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.