missing translation for 'onlineSavingsMsg'
Learn More

D-3-Bromocamphor-8-sulfonic acid ammonium salt, 98%

Product Code. 11355289
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355289 100 g 100g
11335289 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355289 Supplier Thermo Scientific Alfa Aesar Supplier No. A19886.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14575-84-9 | C10H18BrNO4S | 328.221 g/mol

D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.

Solubility
Soluble in Methanol and Water (200 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14575-84-9
Molecular Formula C10H18BrNO4S
Molecular Weight (g/mol) 328.221
MDL Number MFCD00167438
InChI Key GFBVBBRNPGPROZ-ZEMXJPTRSA-N
Synonym d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate
PubChem CID 71306887
IUPAC Name azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N

Specifications

Melting Point ∼284°C (decomposition)
Quantity 100 g
Solubility Information Soluble in Methanol and Water (200g/L) at 25°C.
Formula Weight 328.23
Percent Purity 98%
Chemical Name or Material D-3-Bromocamphor-8-sulfonic acid ammonium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.