missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine methyl ester hydrochloride, 96%, Thermo Scientific Chemicals

Catalog Number 11339505
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
19883-41-1
C9H12ClNO2
201.65
MFCD00137487
DTHMTBUWTGVEFG-DDWIOCJRSA-N
h-d-phg-ome.hcl, h-d-phg-ome hcl, r-methyl 2-amino-2-phenylacetate hydrochloride, r-2-phenylglycine methyl ester hydrochloride, r---2-phenylglycine methyl ester hydrochloride, methyl 2r-amino phenyl acetate hydrochloride, d-alpha-phenylglycine methyl ester hydrochloride, d---2-phenylglycine methyl ester hydrochloride, methyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
12247453
methyl (2R)-2-amino-2-phenylacetate;hydrochloride
COC(=O)C(C1=CC=CC=C1)N.Cl
This item is not returnable. View return policy
19883-41-1
C9H12ClNO2
201.65
MFCD00137487
DTHMTBUWTGVEFG-DDWIOCJRSA-N
h-d-phg-ome.hcl, h-d-phg-ome hcl, r-methyl 2-amino-2-phenylacetate hydrochloride, r-2-phenylglycine methyl ester hydrochloride, r---2-phenylglycine methyl ester hydrochloride, methyl 2r-amino phenyl acetate hydrochloride, d-alpha-phenylglycine methyl ester hydrochloride, d---2-phenylglycine methyl ester hydrochloride, methyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
12247453
methyl (2R)-2-amino-2-phenylacetate;hydrochloride
COC(=O)C(C1=CC=CC=C1)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽190°C (decomposition)
Beilstein 3915654
Formula Weight 201.65
Percent Purity 96%
Quantity 25 g
Optical Rotation −118° (c=1 in Water)
Chemical Name or Material D-(-)-2-Phenylglycine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'