missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%

Product Code. 11381327
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11381327

Brand: Thermo Scientific Alfa Aesar L01564.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17609-48-2 | C10H14ClNO2 | 215.677 g/mol

D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17609-48-2
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.677
MDL Number MFCD00052005
InChI Key FNNXQLSKQSVNLL-SBSPUUFOSA-N
Synonym r-ethyl 2-amino-2-phenylacetate hydrochloride, r-ethyl 2-amino-2-phenylacetate hcl, ethyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine-oet hcl, h-d-phg-oet . hcl, d---2-phenylglycine ethyl ester hydrochloride, d---alpha-phenylglycine ethyl ester hydrochloride, pubchem10931, d-phg-oet hydrochloride
PubChem CID 20833765
IUPAC Name ethyl (2R)-2-amino-2-phenylacetate;hydrochloride
SMILES CCOC(=O)C(C1=CC=CC=C1)N.Cl

Specifications

Melting Point ∽190°C (decomposition)
Beilstein 5775455
Solubility Information Soluble in water.
Quantity 10 g
Formula Weight 215.68
Optical Rotation −94° (c=1 in Water)
Percent Purity ≥98%
Chemical Name or Material D-(-)-2-Phenylglycine ethyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.