missing translation for 'onlineSavingsMsg'
Learn More

D(-)-2-Aminobutyric Acid, 99+%, Thermo Scientific Chemicals

Product Code. 10359273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10359273 25 g Glass bottle 25g
10357610 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10359273 Supplier Thermo Scientific Acros Supplier No. 152140250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-91-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00064414
InChI Key QWCKQJZIFLGMSD-GSVOUGTGSA-N
Synonym d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid
PubChem CID 439691
ChEBI CHEBI:28797
IUPAC Name (2R)-2-aminobutanoic acid
SMILES CC[C@@H](N)C(O)=O

Specifications

Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C2H5CH(NH2)CO2H
Quantity 25 g
Beilstein 04, 408
Specific Rotation Condition −21.20° (20°C c=2,6N HCl)
Specific Rotation -21.2
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol,insoluble in ether
Formula Weight 103.12
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-2-Aminobutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.