missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Aminobutyric acid, 98+%

Product Code. 11368965
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368965 5 g 5g
11358965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368965 Supplier Thermo Scientific Alfa Aesar Supplier No. L14096.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2623-91-8 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-91-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00064414
InChI Key QWCKQJZIFLGMSD-GSVOUGTGSA-N
Synonym d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid
PubChem CID 439691
ChEBI CHEBI:28797
IUPAC Name (2R)-2-aminobutanoic acid
SMILES CC[C@@H](N)C(O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 1720934
Optical Rotation −20.5° (c=4.8 in 6N HCl)
Formula Weight 103.12
Percent Purity ≥98%
Chemical Name or Material D-(-)-2-Aminobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.