missing translation for 'onlineSavingsMsg'
Learn More

D-2-Aminoadipic acid, 98%, Thermo Scientific™

Product Code. 11915391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11915391 5 g 5g
11905391 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11915391

Brand: Thermo Scientific Acros 441960050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7620-28-2
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
InChI Key OYIFNHCXNCRBQI-SCSAIBSYSA-N
Synonym d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
PubChem CID 165627
ChEBI CHEBI:37025
IUPAC Name (2R)-2-aminohexanedioic acid
SMILES C(CC(C(=O)O)N)CC(=O)O

Specifications

Melting Point 186°C to 188°C
Color White
Elemental Analysis 43.60 to 45.84% C, 8.47 to 8.91% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Specific Rotation -20.5
Formula Weight 161.16
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material D-2-Aminoadipic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.