missing translation for 'onlineSavingsMsg'
Learn More

D-2-Aminoadipic acid, 98%, Thermo Scientific™

Product Code. 15443667
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443667 250 mg 250mg
11387089 1 g 1g
11397089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15443667 Supplier Thermo Scientific Alfa Aesar Supplier No. H27345.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7620-28-2
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.157
MDL Number MFCD00063118
InChI Key OYIFNHCXNCRBQI-SCSAIBSYSA-N
Synonym d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
PubChem CID 165627
ChEBI CHEBI:37025
IUPAC Name (2R)-2-aminohexanedioic acid
SMILES C(CC(C(=O)O)N)CC(=O)O

Spécification

Melting Point 205°C
Quantity 250 mg
Beilstein 1724347
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material D-2-Aminoadipic acid
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.