missing translation for 'onlineSavingsMsg'
Learn More

D(+)-2-Amino-3-phenyl-1-propanol, 98%

Product Code. 10357321 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10357321 10 g 10g
10707972 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10357321 Supplier Thermo Scientific Acros Supplier No. 417340100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5267-64-1 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5267-64-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00064298
Synonym d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol

Specifications

Melting Point 90°C to 94°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation + 23
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,benzene,chloroform
Formula Weight 151.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material D(+)-2-Amino-3-phenyl-1-propanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • D(+)-2-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.