missing translation for 'onlineSavingsMsg'
Learn More

D(+)-10-Camphorsulfonyl chloride, 97%

Product Code. 10164590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164590 25 g Glass bottle 25g
10143790 5 g Glass bottle 5g
10636422 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164590 Supplier Thermo Scientific Acros Supplier No. 204630250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21286-54-4 | C10H15ClO3S | 250.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21286-54-4
Molecular Formula C10H15ClO3S
Molecular Weight (g/mol) 250.74
MDL Number MFCD00064156
InChI Key BGABKEVTHIJBIW-GMSGAONNSA-N
Synonym d +-10-camphorsulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride, 1s-+-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, camphor-10-sulfonyl chloride, +-camphor-10-sulfonyl chloride, +-camphor-10-sulfonic acid chloride
PubChem CID 88856
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C

Specifications

Melting Point 63.0°C to 68.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Quantity 25 g
Beilstein 11, 316
Fieser 01,109; 15,68
Specific Rotation + 33.00
Formula Weight 250.74
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-10-Camphorsulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • D(+)-10-Camphorsulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.