Learn More
D(+)-10-Camphorsulfonyl chloride, 97%, Thermo Scientific Chemicals
£24.71 - £314.68
Chemical Identifiers
CAS | 21286-54-4 |
---|---|
Molecular Formula | C10H15ClO3S |
Molecular Weight (g/mol) | 250.74 |
MDL Number | MFCD00064156 |
InChI Key | BGABKEVTHIJBIW-GMSGAONNSA-N |
Synonym | d +-10-camphorsulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride, 1s-+-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, camphor-10-sulfonyl chloride, +-camphor-10-sulfonyl chloride, +-camphor-10-sulfonic acid chloride |
PubChem CID | 88856 |
IUPAC Name | [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride |
SMILES | CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10143790
|
Thermo Scientific Acros
204630050 |
5 g | Glass bottle | ||||||
10164590
|
Thermo Scientific Acros
204630250 |
25 g | Glass bottle | ||||||
10636422
|
Thermo Scientific Acros
204631000 |
100 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21286-54-4 | |
250.74 | |
BGABKEVTHIJBIW-GMSGAONNSA-N | |
88856 | |
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C |
C10H15ClO3S | |
MFCD00064156 | |
d +-10-camphorsulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride, 1s-+-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, camphor-10-sulfonyl chloride, +-camphor-10-sulfonyl chloride, +-camphor-10-sulfonic acid chloride | |
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride |
Specifications
21286-54-4 | |
White to Beige | |
96% min. (ex Cl) (Argentometry) | |
MFCD00064156 | |
01,109; 15,68 | |
d +-10-camphorsulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride, 1s-+-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, camphor-10-sulfonyl chloride, +-camphor-10-sulfonyl chloride, +-camphor-10-sulfonic acid chloride | |
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C | |
250.74 | |
250.74 | |
Crystalline Powder |
63.0°C to 68.0°C | |
Authentic | |
C10H15ClO3S | |
11, 316 | |
+ 33.00 | |
BGABKEVTHIJBIW-GMSGAONNSA-N | |
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride | |
88856 | |
97% | |
D(+)-10-Camphorsulfonyl chloride, 97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-314-8
- D(+)-10-Camphorsulfonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only