missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-[1-13C]Xylose, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | D-Xylose 1-13C |
| CAS | 70849-21-7 |
| Molecular Formula | 13C C4 H10 O5 |
| Quantity | 50 mg |
| Synonym | D-Xylose-1-13C, D-[1-13C]Xylose |
| InChI Formula | InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5+/m0/s1/i1+1 |
| SMILES | OC[C@@H](O)[C@H](O)[C@@H](O)[13CH]=O |
| IUPAC Name | (2R,3S,4R)-2,3,4,5-tetrahydroxy(1^{13}C)pentanal |
| Molecular Weight (g/mol) | 151.12 |
| Formula Weight | 151.0562 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction