missing translation for 'onlineSavingsMsg'
Learn More

CZC24832, MedChemExpress

Product Code. 16469017
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469017 5 mg 5mg
16466428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469017 Supplier Medchem Express Supplier No. HY15294
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CZC24832 is a highly selective and potent PI3Kγ inhibitor (IC50=27 nM) with apparent dissociation constants (Kdapp) of 19 nM.

CZC24832 is a highly selective and potent PI3Kγ inhibitor (IC50=27 nM) with apparent dissociation constants (Kdapp) of 19 nM.

Specifications

Chemical Name or Material CZC24832
CAS 1159824-67-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H17FN6O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 53 mg/mL (145.44 mM)
SMILES FC1=CC(C2=CC(S(=O)(NC(C)(C)C)=O)=CN=C2)=CN3C1=NC(N)=N3
Molecular Weight (g/mol) 364.4
Formula Weight 364.4
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.