missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYYR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580536
This item is not returnable.
View return policy
Description
This antibody has specificity for Human CYYR1.
CYYR1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
Specifications
| CYYR1 | |
| Polyclonal | |
| Unconjugated | |
| CYYR1 | |
| C21orf95; cysteine and tyrosine rich 1; cysteine and tyrosine-rich 1; cysteine and tyrosine-rich 1 protein; cysteine and tyrosine-rich protein 1; cysteine and tyrosine-rich protein 1 isoform 1,2,2bis,3,4; cysteine and tyrosine-rich protein 1 isoform 1,2,3,4b; cysteine and tyrosine-rich protein 1 isoform 1,2,3b; cysteine and tyrosine-rich protein 1 isoform 1,2,4; cysteine/tyrosine-rich 1; Cyyr1; hypothetical protein zgc:77252; LOW QUALITY PROTEIN: cysteine and tyrosine-rich protein 1; proline-rich domain-containing protein; zgc:77252 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96J86 | |
| CYYR1 | |
| A synthetic peptide corresponding to the C-terminus of the Human CYYR1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction