missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTSB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13851-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the cytospin-A family. It is localized in the nucleus, and highly expressed in testis and some cancer cell lines. A chromosomal translocation involving this gene and platelet-derived growth factor receptor, beta gene (PDGFRB) may be a cause of juvenile myelomonocytic leukemia. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| CYTSB | |
| Polyclonal | |
| Unconjugated | |
| SPECC1 | |
| cytospin B, CYTSB, HCMOGT 1, NSP, NSP5, Nuclear structure protein 5, SPECC1, Sperm antigen HCMOGT 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303208, 432572, 92521 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5M775, Q5SXY1 | |
| Specc1 | |
| CYTSB Fusion Protein Ag4822 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction