missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTSA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25390-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is leucine-rich and is thought to play a role in regulating the interaction of the cytoskeleton with a variety of cellular processes. Mutations in this gene are associated with the French-Canadian type of Leigh syndrome. Transcripts ranging in size from 4.8 to 7.0 kb which result from alternative polyadenylation have been reported for this gene.Specifications
| CYTSA | |
| Polyclonal | |
| Unconjugated | |
| SPECC1L | |
| cytospin A, CYTSA, KIAA0376, SPECC1 like protein, SPECC1L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23384, 74392 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q2KN98, Q69YQ0 | |
| Specc1l | |
| CYTSA Fusion Protein Ag21896 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction