missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CytoVista™ 10X Wash Buffer

Product Code. 16369563
Click to view available options
Quantity:
70 mL
Unit Size:
70mL
This item is not returnable. View return policy

Product Code. 16369563

Brand: Invitrogen™ V11311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CytoVista 10X Wash Buffer is used in wash steps when performing immunohistochemistry (IHC) or immuno-labeling with tissue or 3D cell culture.

This buffer is 10X concentrated and should be diluted to 1X with PBS before use. It consists of PBS with 0.2% Tween-20 and 10 μg/mL heparin and helps enable sharp and bright images at any depth of tissue or 3D cell culture when used with other CytoVista products.

Features of the CytoVista family of tissue and 3D cell culture clearing reagents include:
• Rapid clearing of fluorescently labeled tissue for 3D imaging
• Minimal shrinkage, expansion, or other morphological changes to tissue
• Simple workflow does not require any special instruments or equipment and is easily adapted to IHC
• Compatible with most fluorophores, including dyes and fluorescent proteins

The CytoVista tissue and 3D cell culture clearing workflow clears tissue or 3D cultured cells with minimal changes to morphology or the sensitivity of detection with almost any fluorophore. With an easy-to-use protocol, whole mouse brain (8-mm section) can be cleared in 24 hours, or a 1-mm section in 2 hours, without the use of any special instruments or equipment.

The CytoVista clearing workflow is compatible with most fluorophores, including fluorescent proteins that are detected with common fluorescent imaging instruments, such as wide-field microscopes, high content analyzers, confocal microscopes, and lightsheet microscopes. For first time users, we recommend the CytoVista Tissue Clearing/Staining Kit (Cat No V11324) or CytoVista 3D Cell Culture Clearing/Staining Kit (Cat No. V11325). These kits contain all necessary reagents for both clearing and IHC staining.

CytoVista clearing technology sufficiently clears tissue for 3D fluorescent imaging, but unlike other clearing techniques does not significantly changes the tissue morphology. Minimal morphological changes, such as shrinkage and contraction, have been observed. For precious samples, the clearing can be reversed, and the tissue can be processed for histology studies, like H&E staining.

For tissues that are easier to clear, like brain, lung, intestine, skin, and tissues of less than 250-μm thickness, treatment with CytoVista Tissue Clearing Reagent is sufficient for 3D imaging. However, tissues of more than 250-μm thickness or that are hard to clear like kidney, liver, heart, and placenta will require an initial treatment with CytoVista Tissue Clearing Reagent followed by treatment with CytoVista Tissue Clearing Enhancer.

CytoVista 3D Cell Culture Clearing Reagent is recommended for spheroids or organoids up to 1 mm in thickness.

For hard tissues like bone, cartilage, and connective tissue, CytoVista Tissue Clearing Reagent is not recommended.

TRUSTED_SUSTAINABILITY

Specifications

Type Wash Buffer
Product Line CytoVista
Reagent Type Wash Buffer
Content And Storage Store in refrigerator (2°C to 8°C).Use sterile techniques when using. If necessary, sterile-filter through 0.22-μm filter before each use.
Quantity 70 mL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.