missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cytosporone B

Description
Cytosporone B is a naturally occurring Nur77 (NR4A1) agonist (Kd = 8.52 x 10-7 M). Induces apoptosis in gastric cancer cells in vitro. Increases blood glucose levels in fasting C57 mice and suppresses xenograft tumor growth. Also exhibits a potent antifibrotic effect in bleomycin-induced skin fibrosis in vivo.
Specifications
Specifications
| Chemical Name or Material | Ethyl 3,5-dihydroxy-2-(1-oxooctyl)-benzeneacetate |
| CAS | 321661-62-5 |
| Quantity | 50 mg |
| Target | Nuclear Receptor Agonists |
| Molecular Formula | C18H26O5 |
| Formula Weight | 322.4 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?