missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 4A1/SULT4A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81770
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 4A1/SULT4A1 Polyclonal specifically detects Cytosolic Sulfotransferase 4A1/SULT4A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytosolic Sulfotransferase 4A1/SULT4A1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Brain sulfotransferase-like protein, BRSTL1, BR-STL-1, EC 2.8.2.-, hBR-STL, hBR-STL-1MGC40032, nervous system cytosolic sulfotransferase, Nervous system sulfotransferase, NST, ST4A1, sulfotransferase 4A1, sulfotransferase family 4A, member 1, sulfotransferase-related protein, SULTX3DJ388M5.3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25830 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SULT4A1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GEFESKYFEFHGVRLPPFCRGKMEEIANFPVRPSDVWIVTYPKSGTSLLQEVVYLVSQGADPDEIGLMNIDEQLPVLEYPQPGLDIIKELTSPRLIKSHLPYRFLPSDLHNGDSKVIYMARNPKDLVVSYYQFHRSL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction