missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 2B1/SULT2B1 Antibody (007), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18335141 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335141 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335141 Supplier Novus Biologicals Supplier No. NBP289895AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytosolic Sulfotransferase 2B1/SULT2B1 Monoclonal specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 2B1/SULT2B1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 2B1/SULT2B1 (NP_004596.2; Asp 2-Ser 365) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6820
Test Specificity No cross-reactivity in ELISA with: Human SULT2A1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.