missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 2A1/SULT2A1 Rabbit anti-Human, Rat, Clone: 4A3H6, Novus Biologicals™

Product Code. 18363513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363513 100 μg 100µL
18390844 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18363513 Supplier Novus Biologicals Supplier No. NBP316665100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cytosolic Sulfotransferase 2A1/SULT2A1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 2A1/SULT2A1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 4A3H6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-285 of human Cytosolic Sulfotransferase 2A1/SULT2A1 (Q06520).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.